Learn More
GRANT INSTRUMENTS Microcentrifuge/vortex mixer Combi-spin™ to 3500 rpm, inc. rotors for 12 x 1.5ml and 12 x 0.5ml plus 12 x 0.2ml
SDP
Catalog No. NC1655892
Encompass_Preferred
The Centrifuge/Vortexer Combi-spin PCV-2400 is specially designed for life science research. It can be used in biochemical, microbiological and industrial biotechnology laboratories.A cost-effective fixed-speed combined centrifuge/vortex mixer for combined or independent centrifuge and mixing applications of microtubes and 0.2 ml microtube strips in low volume applications.Centrifugation 2800rpm at 50Hz and 3500rpm at 60Hz.The vortex cup emerges through an opening in the closed lid, enabling individual tubes to be mixed while a full load of tubes is spinning.Very simple to operate - use the left-hand button to select continuous or short spin. A safety interlock stops the rotor when the lid is opened to ensure you and your workplace remain safe.The Combi-spin is supplied as standard with two interchangeable rotors for 12x1.5ml and 12x0.5ml + 12x0.2ml microtubes.
Catalog No. NC1655892 Supplier GRANT INSTRUMENTS Supplier No. PCV2400
The Centrifuge/Vortexer Combi-spin PCV-2400 is specially designed for life science research. It can be used in biochemical, microbiological and industrial biotechnology laboratories.A cost-effective fixed-speed combined centrifuge/vortex mixer for combined or independent centrifuge and mixing applications of microtubes and 0.2 ml microtube strips in low volume applications.Centrifugation 2800rpm at 50Hz and 3500rpm at 60Hz.The vortex cup emerges through an opening in the closed lid, enabling individual tubes to be mixed while a full load of tubes is spinning.Very simple to operate - use the left-hand button to select continuous or short spin. A safety interlock stops the rotor when the lid is opened to ensure you and your workplace remain safe.The Combi-spin is supplied as standard with two interchangeable rotors for 12x1.5ml and 12x0.5ml + 12x0.2ml microtubes.