Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CombiMethanol Solvent, MilliporeSigma™

Catalog No. M1880091000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1880091000 1 L
M1880092500 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1880091000 Supplier MilliporeSigma Supplier No. 1.88009.1000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CombiMethanol Solvent for volumetric Karl Fischer titration with one component reagents max. 0.01% H2O Aquastar™.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methanol
CAS 67-56-1
Melting Point −98°C
Boiling Point 64.5°C (1013 hPa)
Density 0.792 g/cm3 (at 20°C)
Quantity 1 L
Flash Point 9.7°C
Molecular Formula CH4O
Packaging Glass Bottle
Synonym MeOH, Hydroxymethane, Methyl alcohol, Carbinol, Hydroxymethane, Methyl alcohol, Carbinol
Molecular Weight (g/mol) 32.04
Grade Karl Fischer
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.