Learn More
Dentec Safety™ Combination Gas Cartridge
SDP
Catalog No. 19222526 Shop All Dentec Safety Specialists Inc. Products
Change view
Click to view available options
Filter Type:
Combination Acid Gases and P100
Combination Acid Gases/Organic Vapor and P100
Combination Ammonia/Methylamine and P100
Combination Multi-Vapor Gas and P100
Combination Organic Vapor and P100
Quantity:
4/Box
60/Box
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Filter Type Quantity
19-222-526 Combination Multi-Vapor Gas and P100 60/Box
19-222-519 Combination Acid Gases and P100 4/Box
19-222-520 Combination Acid Gases and P100 60/Box
19-222-523 Combination Acid Gases/Organic Vapor and P100 4/Box
19-222-524 Combination Acid Gases/Organic Vapor and P100 60/Box
19-222-521 Combination Ammonia/Methylamine and P100 4/Box
19-222-522 Combination Ammonia/Methylamine and P100 60/Box
19-222-525 Combination Multi-Vapor Gas and P100 4/Box
19-222-517 Combination Organic Vapor and P100 4/Box
19-222-518 Combination Organic Vapor and P100 60/Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 19-222-526 Supplier Dentec Safety™ Supplier No. 158T27LP0B60
Add to Cart
Add to Cart
Dentec Safety™ Combination Gase Cartridge features unique space plenums that allow users to access the entire filter material reducing breathing resistance, providing longer cartrdige life, reducing costs and improving productivity
  • Offset connector allows the user to rotate the cartridge into 3 different positions improving visibility
  • Saf-Twist design quickly attaches the cartridge to any Dentec Safety half-mask or full face air-purifying respirator Facepiece.
Specifications
Filter Type Combination Multi-Vapor Gas and P100
Material Plastic
Product Type Respirator Accessories
For Use With (Equipment) Masks
Includes Cartridges and filters
Quantity 60/Box