Learn More
Waters Corp Waters Corporation Autosampler Vial, Polypropylene, 12 x 32 mm, 300 μL
Catalog No. 50782173 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This clean polypropylene-molded vial is preferred in applications where there is a concern that a compound or molecule may stick to a polar glass surface. It offers a cost-effective alternative to glass vials.
  • Constructed from polypropylene
  • Measures 12 x 32 mm
  • Offers a volume capacity of 300 μL
  • Features a screw neck closure type
  • Includes a polyethylene septumless cap
  • Designed with a conical format
  • Supplied with vial, cap, & septum
  • Suitable for autosampler applications
  • Provides a cost-effective alternative to glass vials
  • Ideal for sensitive applications where compounds may adhere to glass surfaces
Catalog No. 50-782-173 Supplier Waters Corp Supplier No. 186004112
This clean polypropylene-molded vial is preferred in applications where there is a concern that a compound or molecule may stick to a polar glass surface. It offers a cost-effective alternative to glass vials.
  • Constructed from polypropylene
  • Measures 12 x 32 mm
  • Offers a volume capacity of 300 μL
  • Features a screw neck closure type
  • Includes a polyethylene septumless cap
  • Designed with a conical format
  • Supplied with vial, cap, & septum
  • Suitable for autosampler applications
  • Provides a cost-effective alternative to glass vials
  • Ideal for sensitive applications where compounds may adhere to glass surfaces