Learn More
Waters Corp LCGC Certified Max Recovery Vial, Clear Glass, 2 mL, 12 x 32 mm, Screw Neck
Catalog No. 50782509 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These certified vials are designed for critical analytical applications, ensuring safe and secure sample storage without spillage. They are tested for cleanliness by HPLC and held to the tightest dimensional tolerances, providing an inert and compositionally stable solution for various laboratory needs.
  • Certified by HPLC for cleanliness and tight dimensional tolerances.
  • Ensures safe and secure sample storage without spillage.
  • Inert composition prevents interference in analysis.
  • Withstands chemical and mechanical forces for reliable use.
  • Thoroughly tested throughout manufacturing and after packaging.
  • Ideal for mass spectrometry (MS) applications.
  • Excellent chemical compatibility and solvent resistance.
  • Screw cap design provides a secure seal.
Catalog No. 50-782-509 Supplier Waters Corp Supplier No. 186000326C
These certified vials are designed for critical analytical applications, ensuring safe and secure sample storage without spillage. They are tested for cleanliness by HPLC and held to the tightest dimensional tolerances, providing an inert and compositionally stable solution for various laboratory needs.
  • Certified by HPLC for cleanliness and tight dimensional tolerances.
  • Ensures safe and secure sample storage without spillage.
  • Inert composition prevents interference in analysis.
  • Withstands chemical and mechanical forces for reliable use.
  • Thoroughly tested throughout manufacturing and after packaging.
  • Ideal for mass spectrometry (MS) applications.
  • Excellent chemical compatibility and solvent resistance.
  • Screw cap design provides a secure seal.