Learn More
Waters Corp Waters Max Recovery Vial, LCGC Certified, Clear Glass, 12 x 32 mm, 2 mL Volume
Catalog No. 50782560 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters Certified vials are tested for cleanliness by HPLC and held to the tightest dimensional tolerances in the industry. The LCGC Certified Clear Glass Vial is a 12 x 32 mm autosampler vial, a polyethylene and septumless product that works seamlessly with your chosen autosampler. These vials are strictly tested for cleanliness to ensure that only the cleanest vials are received, reducing the risk of sample contamination.
  • Tested for cleanliness by HPLC
  • Manufactured to tight dimensional tolerances
  • Designed for seamless integration with autosamplers
  • Features a polyethylene and septumless cap
  • Max recovery vial format
  • Reduces the risk of sample contamination
Catalog No. 50-782-560 Supplier Waters Corp Supplier No. 186004168C
Waters Certified vials are tested for cleanliness by HPLC and held to the tightest dimensional tolerances in the industry. The LCGC Certified Clear Glass Vial is a 12 x 32 mm autosampler vial, a polyethylene and septumless product that works seamlessly with your chosen autosampler. These vials are strictly tested for cleanliness to ensure that only the cleanest vials are received, reducing the risk of sample contamination.
  • Tested for cleanliness by HPLC
  • Manufactured to tight dimensional tolerances
  • Designed for seamless integration with autosamplers
  • Features a polyethylene and septumless cap
  • Max recovery vial format
  • Reduces the risk of sample contamination