Learn More
Waters Corp Waters LCMS Certified Clear Glass Vial, 12 x 32 mm, 2 mL, Screw Neck
Catalog No. 50782561 Shop All Waters Corporation Products
Encompass_Preferred
These LCMS Certified vials are manufactured under tightly controlled processes and handling procedures. Each lot is tested with mass spectrometers and includes a certificate of analysis, ensuring suitability for LCMS analysis. They come with a cap and are made from high-quality glass, designed to withstand high temperatures and provide secure long-term sample storage.
  • Manufactured following tightly controlled processes and handling procedures.
  • Tested with mass spectrometers and packaged with a certificate of analysis.
  • Features a screw neck closure for secure sealing.
  • Cap color is blue.
  • Seal material is preslit PTFE/Silicone.
  • Volume capacity is 2 mL.
  • Dimensions are 12 x 32 mm.
  • Suitable for sample preparation applications.
Catalog No. 50-782-561 Supplier Waters Corp Supplier No. 600000668CV
These LCMS Certified vials are manufactured under tightly controlled processes and handling procedures. Each lot is tested with mass spectrometers and includes a certificate of analysis, ensuring suitability for LCMS analysis. They come with a cap and are made from high-quality glass, designed to withstand high temperatures and provide secure long-term sample storage.
  • Manufactured following tightly controlled processes and handling procedures.
  • Tested with mass spectrometers and packaged with a certificate of analysis.
  • Features a screw neck closure for secure sealing.
  • Cap color is blue.
  • Seal material is preslit PTFE/Silicone.
  • Volume capacity is 2 mL.
  • Dimensions are 12 x 32 mm.
  • Suitable for sample preparation applications.