Learn More
Waters Corp Vial, LCMS Certified, Clear Glass, 12 x 32 mm, 2 mL
Catalog No. 50782565 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These LCMS Certified vials are manufactured using tightly controlled processes and handling procedures. Each vial is tested with mass spectrometers and includes a certificate of analysis detailing the reference and vial scan for the manufacturing lot. Designed for use with autosamplers, these vials facilitate sample preparation at scale, meeting high standards of dimensional and cleanliness tolerances to prevent contamination and ensure data integrity. The included cap and septum enable clean and easy sample injection.
  • LCMS certified
  • Screw neck closure
  • Blue cap
  • 12 × 32 mm dimensions
  • Max recovery vial format
  • Clear glass material
  • PTFE/Silicone seal material
  • 1.5 mL volume capacity
  • Autosampler vial type
  • Includes vial, cap, & septum
  • Sample limited
  • Designed for sample preparation
  • 100 units per package
Catalog No. 50-782-565 Supplier Waters Corp Supplier No. 600000749CV
These LCMS Certified vials are manufactured using tightly controlled processes and handling procedures. Each vial is tested with mass spectrometers and includes a certificate of analysis detailing the reference and vial scan for the manufacturing lot. Designed for use with autosamplers, these vials facilitate sample preparation at scale, meeting high standards of dimensional and cleanliness tolerances to prevent contamination and ensure data integrity. The included cap and septum enable clean and easy sample injection.
  • LCMS certified
  • Screw neck closure
  • Blue cap
  • 12 × 32 mm dimensions
  • Max recovery vial format
  • Clear glass material
  • PTFE/Silicone seal material
  • 1.5 mL volume capacity
  • Autosampler vial type
  • Includes vial, cap, & septum
  • Sample limited
  • Designed for sample preparation
  • 100 units per package