Learn More
World Precision Instrument Combination pH Electrode, low noise
SDP
Catalog No. 502538633
Encompass
WPI's pH-Silver, single/double-junction, gel-sealed Ag/AgCl combination pH electrodes are very low noise. They are designed for both laboratory and field use, especailly for applications involving strong magnetic fields. The unique feature of these electrodes provide unparalleled performance.
Catalog No. 50-253-8633 Supplier World Precision Instrument Supplier No. PHSILVER
WPI's pH-Silver, single/double-junction, gel-sealed Ag/AgCl combination pH electrodes are very low noise. They are designed for both laboratory and field use, especailly for applications involving strong magnetic fields. The unique feature of these electrodes provide unparalleled performance.