Learn More
Chemglass Life Sciences Combination Spatula, Spoon and Scoop, Stainless Steel
SDP
Catalog No. 501311188
Encompass_Preferred
Combination Spatula, Spoon and Scoop, Stainless Steel. Made entirely of Stainless Steel, this spatula has both a spoon and scoop. The spoon has a diameter of 26mm while the overall length of the spatula is 195mm. Sold 6 per package.
Catalog No. 50-131-1188 Supplier Chemglass Life Sciences Supplier No. CG197980
Combination Spatula, Spoon and Scoop, Stainless Steel. Made entirely of Stainless Steel, this spatula has both a spoon and scoop. The spoon has a diameter of 26mm while the overall length of the spatula is 195mm. Sold 6 per package.