Learn More
MSC PEC Tools Combination Square Set, 24 in., Cast Iron, 1 ea.
Catalog No. 5000515668
Encompass
This 2-piece combination square set provides essential tools for precise layout and measurement in metalworking applications. Constructed with high-quality components, the set includes a 24 in. (600 mm) blade and a versatile square/miter head.
  • Features fully hardened and tempered blades for maximum durability
  • Includes a satin chrome finish on blades to reduce glare and improve readability
  • Provides high contrast, micro-fine graduations for increased measurement accuracy
  • Equipped with a level vial and a hardened scribe tool built into the square/miter head
  • Supplied with a durable hammertone powder-coat finish on the heads
  • Offers E/M graduation styles including 0.5 mm and 32nds, and mm and 64ths
Catalog No. 50-005-15668 Supplier MSC Supplier No. 83305920
This 2-piece combination square set provides essential tools for precise layout and measurement in metalworking applications. Constructed with high-quality components, the set includes a 24 in. (600 mm) blade and a versatile square/miter head.
  • Features fully hardened and tempered blades for maximum durability
  • Includes a satin chrome finish on blades to reduce glare and improve readability
  • Provides high contrast, micro-fine graduations for increased measurement accuracy
  • Equipped with a level vial and a hardened scribe tool built into the square/miter head
  • Supplied with a durable hammertone powder-coat finish on the heads
  • Offers E/M graduation styles including 0.5 mm and 32nds, and mm and 64ths