Learn More
MSC PEC Tools Combination Square Set, Cast Iron, 6 in., English/Metric Graduation, 1 ea
Catalog No. 5000514971
Encompass
This two-piece 15.24 cm (6 in.) combination square set is a versatile measuring tool designed for precision layout and inspection tasks. The set includes a durable blade and a square/miter head, both constructed from cast iron for stability and long-lasting performance.
  • Features hardened and tempered blades for durability
  • Includes satin chrome finish for improved readability
  • Provides high-contrast, micro-fine blade graduations in both English and metric scales
  • Includes square/miter head equipped with a level vial and a hardened scribe tool
  • Features a hammertone powder-coat finish on the heads
Catalog No. 50-005-14971 Supplier MSC Supplier No. 83305896
This two-piece 15.24 cm (6 in.) combination square set is a versatile measuring tool designed for precision layout and inspection tasks. The set includes a durable blade and a square/miter head, both constructed from cast iron for stability and long-lasting performance.
  • Features hardened and tempered blades for durability
  • Includes satin chrome finish for improved readability
  • Provides high-contrast, micro-fine blade graduations in both English and metric scales
  • Includes square/miter head equipped with a level vial and a hardened scribe tool
  • Features a hammertone powder-coat finish on the heads