Learn More
Grainger Combination Wye 1/8 Bend SS 4 x 4 x 2 in
Catalog No. 502751805
Encompass_Preferred
The Blucher 14N994 is a stainless steel reducing wye fitting with female NPT threads. It reduces from 1-1/2" to 1" to 3/4". The body is made from 316 stainless steel for corrosion resistance. Maximum pressure is 300 psi at 70°F. Temperature range is -20°F to 250°F. Complies with MSS SP-114 standards. Made in the USA.
Catalog No. 50-275-1805 Supplier Grainger Supplier No. 14N994
The Blucher 14N994 is a stainless steel reducing wye fitting with female NPT threads. It reduces from 1-1/2" to 1" to 3/4". The body is made from 316 stainless steel for corrosion resistance. Maximum pressure is 300 psi at 70°F. Temperature range is -20°F to 250°F. Complies with MSS SP-114 standards. Made in the USA.