Learn More
Grainger Combination Wye and 1/8 Bend SS 2 in
Catalog No. 502749863
Encompass_Preferred
The Grainger 14N992 is a 14-inch diameter, 992 CFM, direct drive upblast roof exhaust fan. It features a 1/6 HP, 115V, 1-phase, 60Hz motor with permanently lubricated ball bearings and automatic thermal overload protection. The housing is constructed of heavy gauge aluminum with a spun aluminum venturi for maximum airflow. It has a built-in backdraft damper and is ETL listed. This exhaust fan is ideal for ventilating hot and greasy air in commercial kitchens.
Catalog No. 50-274-9863 Supplier Grainger Supplier No. 14N992
The Grainger 14N992 is a 14-inch diameter, 992 CFM, direct drive upblast roof exhaust fan. It features a 1/6 HP, 115V, 1-phase, 60Hz motor with permanently lubricated ball bearings and automatic thermal overload protection. The housing is constructed of heavy gauge aluminum with a spun aluminum venturi for maximum airflow. It has a built-in backdraft damper and is ETL listed. This exhaust fan is ideal for ventilating hot and greasy air in commercial kitchens.