Learn More
Anton Paar Combine with DMA density meter for dynamic and kinematic viscosity in one cycle. Referenced in USP 913 and Ph. Eur. 2.2.49. Wort compliant (MEBAK, ASBC). Optional sample changer and integration with refractive index and pH.
Catalog No. 5000580537
Encompass_Preferred
MEASURING RANGE Dynamic viscosity: 0.2 mPa·s to 10,000 mPa·sShear rate: 0.5 s-1 to 1,000 s-1Temperature; 5 °C to 100 °C (41 °F to 212 °F)-40 °C to +100 °C (-40 °F to +212 °F) (with counter cooling)1)PRECISION Viscosity: Repeatability s.d.: 0.1 % 2)Viscosity: Accuracy: 0.5 % 3)Measuring time: Resolution: 0.001 sMeasuring time: Accuracy: 0.05 %Temperature: Repeatability s.d.: 0.005 °CTemperature: Accuracy: 0.02 °CInclination: Repeatability s.d.: 0.02 °Inclination: Accuracy: 0.1 °
Catalog No. 50-005-80537 Supplier Anton Paar Supplier No. 314856
Please contact your sales representative for a quote or further ordering instructions.
MEASURING RANGE Dynamic viscosity: 0.2 mPa·s to 10,000 mPa·sShear rate: 0.5 s-1 to 1,000 s-1Temperature; 5 °C to 100 °C (41 °F to 212 °F)-40 °C to +100 °C (-40 °F to +212 °F) (with counter cooling)1)PRECISION Viscosity: Repeatability s.d.: 0.1 % 2)Viscosity: Accuracy: 0.5 % 3)Measuring time: Resolution: 0.001 sMeasuring time: Accuracy: 0.05 %Temperature: Repeatability s.d.: 0.005 °CTemperature: Accuracy: 0.02 °CInclination: Repeatability s.d.: 0.02 °Inclination: Accuracy: 0.1 °