Learn More
Med Vet International White Abdominal Pad Sterile 7-1/2 X 8 inch 4252, 12/Box
Catalog No. 502829879
Encompass_Preferred
Clean, white color Sealed edges to prevent leaking Superior absorption with virtually no linting Faster wicking for drier wound sites Single use Combine ABD pads have a non-woven outer fabric with inner cellulose tissue construction that helps provide maximum moisture barrier. - Clean, white color. Sealed edges to prevent leaking. Superior absorption with virtually no linting. Faster wicking for drier wound sites. Single use.
Catalog No. 50-282-9879 Supplier Med Vet International Supplier No. ABD8X75STRLBX
Clean, white color Sealed edges to prevent leaking Superior absorption with virtually no linting Faster wicking for drier wound sites Single use Combine ABD pads have a non-woven outer fabric with inner cellulose tissue construction that helps provide maximum moisture barrier. - Clean, white color. Sealed edges to prevent leaking. Superior absorption with virtually no linting. Faster wicking for drier wound sites. Single use.