Learn More
Med Vet International Abdominal (Abd) Pads, 5in. X 9in., Sterile, 25/Box
Catalog No. 502832183
Encompass_Preferred
Medline's sterile abdominal pad has extra absorbency and wicking to handle heavily draining wounds. It has a soft, nonwoven outer layer that quickly wicks fluid to a cellulose center to keep the wound dry. This ABD pad has a hydrophobic back that helps prevent strike-through and sealed edges to prevent linting. Specifications: 5in by 9in.
Catalog No. 50-283-2183 Supplier Med Vet International Supplier No. NON21450H
Medline's sterile abdominal pad has extra absorbency and wicking to handle heavily draining wounds. It has a soft, nonwoven outer layer that quickly wicks fluid to a cellulose center to keep the wound dry. This ABD pad has a hydrophobic back that helps prevent strike-through and sealed edges to prevent linting. Specifications: 5in by 9in.