Learn More
Med Vet International Abdominal Abd (Combine) Pad, 5" X 9", Sterile, Dukal, 25/Box, 400/Case
Catalog No. 502831441
Encompass_Preferred
Super absorbent abdominal pads feature a soft non-woven outer layer that quickly wicks fluid to a cellulose center. This thick layer of cellulose quickly absorbs and disperses fluids laterally to prevent pooling. Hydrophobic back impedes strike through and all edges are sealed to prevent linting. Sterile, 400/Case.
Catalog No. 50-283-1441 Supplier Med Vet International Supplier No. 5590
Super absorbent abdominal pads feature a soft non-woven outer layer that quickly wicks fluid to a cellulose center. This thick layer of cellulose quickly absorbs and disperses fluids laterally to prevent pooling. Hydrophobic back impedes strike through and all edges are sealed to prevent linting. Sterile, 400/Case.