Learn More
Med Vet International Abdominal Abd Combine Dressing, 8 X 10in, Non-Sterile, Dukal, 400/Case
Catalog No. 502830944
Encompass_Preferred
Combine pads consist of three key layers: outer nonwoven layer to wick, fluff filler to absorb and disperse fluid and wet proof moisture barrier to retard fluid strike-through. These pads act as a reservoir, reduces pooling and protects against contamination. 8 x 10in non-sterile, 400/case
Catalog No. 50-283-0944 Supplier Med Vet International Supplier No. 5943
Combine pads consist of three key layers: outer nonwoven layer to wick, fluff filler to absorb and disperse fluid and wet proof moisture barrier to retard fluid strike-through. These pads act as a reservoir, reduces pooling and protects against contamination. 8 x 10in non-sterile, 400/case