Learn More
Med Vet International Abdominal Combine Pad, 8"X10", Sterile, 18/Box
Catalog No. 502830359
Encompass_Preferred
Super absorbent abdominal pads feature a soft non-woven outer layer that quickly wicks fluid to a cellulose center. This thick layer of cellulose quickly absorbs and disperses fluids laterally to prevent pooling. Hydrophobic back impedes strike through and all edges are sealed to prevent linting.
Catalog No. 50-283-0359 Supplier Med Vet International Supplier No. ABD8X10STRL18BX
Super absorbent abdominal pads feature a soft non-woven outer layer that quickly wicks fluid to a cellulose center. This thick layer of cellulose quickly absorbs and disperses fluids laterally to prevent pooling. Hydrophobic back impedes strike through and all edges are sealed to prevent linting.