Learn More
Med Vet International Combine Roll 8" X 20 Yards, Each
Supplier:  Med Vet International 3001
Encompass_Preferred
Combine rolls, or composite padding, is a post-operative dressing used when excessive drainage from wounds demands more absorbency. The absorbent cotton, with a center layer of cellulose, has an exterior wrapping of soft non-woven fabric. Non-Sterile 20 yards in length.
Catalog No. 50-283-1262