Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Combined Quality Control Standard, various concentrations in water with dilute nitric acid, Ultra Scientific

Catalog No. usiqc026
Encompass
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USIQC026 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USIQC026 Supplier Agilent Technologies Supplier No. IQC026
Only null left
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Combined Quality Control Standard
CAS Max % 94.6, 5.0, 0.01, 0.01, 0.01, 0.01
CAS 7429-90-5, 7440-39-3, 7440-42-8, 7440-09-7, 7732-18-5
Analyte or Component Names Aluminum,barium,boron
Element Al, Ba, B
Solvent or Matrix Water/Dilute Nitric Acid
Quantity 125 mL
Concentration or Composition (by Analyte or Components) Various concentrations
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.