Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CombiSolvent Keto, MilliporeSigma™

Catalog No. M1880071002 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1880071002 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1880071002 Supplier MilliporeSigma Supplier No. 1.88007.1002
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

One component solvent reagent specifically formulated for volumetric Karl Fischer titration of aldehydes and ketones.

TRUSTED_SUSTAINABILITY

Specifications

Density 0.924 g/cm3 (at 20°C)
Quantity 1 L
Flash Point 31°C
Packaging Glass Bottle
Grade Karl Fischer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.