Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CombiSolvent, Methanol-free Solvent for Volumetric Karl Fischer Titration, Apura™, MilliporeSigma™

Catalog No. M1880082500 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1880082500 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1880082500 Supplier MilliporeSigma Supplier No. 1.88008.2500
Add to Cart
Add to Cart

Specifications

Chemical Name or Material CombiSolvent, Methanol-free Solvent
Physical Form Liquid
Quantity 2.5 L
Packaging Glass bottle
UN Number UN 1993
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.