Learn More
USA Scientific Inc Combitips advanced, , 0.1mL
Catalog No. 502745008
Encompass
Combitips advanced positive displacement tips, 0.1 mL, white color ring, non-sterile Eppendorf Quality, 4 bags of 25 (100) tips, in dispenser box
Catalog No. 50-274-5008 Supplier USA Scientific Inc Supplier No. 40089405
Combitips advanced positive displacement tips, 0.1 mL, white color ring, non-sterile Eppendorf Quality, 4 bags of 25 (100) tips, in dispenser box