Learn More
USA Scientific Inc Combitips advanced, , 5mL
Catalog No. 502744838
Encompass
Combitips advanced positive displacement tips, 5 mL, blue color ring, non-sterile Eppendorf Quality, 4 bags of 25 (100) tips, in dispenser box
Catalog No. 50-274-4838 Supplier USA Scientific Inc Supplier No. 40089456
Combitips advanced positive displacement tips, 5 mL, blue color ring, non-sterile Eppendorf Quality, 4 bags of 25 (100) tips, in dispenser box