Learn More
USA Scientific Inc Combitips advanced, str, 10mL
Catalog No. 502744844
Encompass
Combitips advanced positive displacement tips, 10 mL, str, individually wrapped, 100/box
Catalog No. 50-274-4844 Supplier USA Scientific Inc Supplier No. 40089570
Combitips advanced positive displacement tips, 10 mL, str, individually wrapped, 100/box