Learn More
USA Scientific Inc Combitips advanced, str, 2.5mL
Catalog No. 502744841
Encompass
Combitips advanced positive displacement tips, 2.5 mL, str, individually wrapped, 100/box
Catalog No. 50-274-4841 Supplier USA Scientific Inc Supplier No. 40089553
Combitips advanced positive displacement tips, 2.5 mL, str, individually wrapped, 100/box