Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CombiTitrant 2, AQUASTAR™, EMD Chemicals

Catalog No. M1880021045 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1880021045 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. M1880021045 Supplier MilliporeSigma Supplier No. 1.88002.1045
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

One component titrant for volumetric Karl Fischer titration with integrated RFID tag for seamless data transfer.

TRUSTED_SUSTAINABILITY

Specifications

CAS 693-98-1
Color Red to Brown
Density 1.090-1.110g/cm3
Physical Form Liquid
Quantity 1 L
Packaging GL45 Poly Coated Glass Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.