Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CombiTitrant 5, Aquastar™, MilliporeSigma™

Catalog No. M1880051000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1880051000 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. M1880051000 Supplier MilliporeSigma Supplier No. 1.88005.1000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

One-component reagent for volumetric Karl Fischer titration.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula Mixture
Synonym carl fischer

Specifications

Color Red to Brown
Density 1.160-1.190g/3
Physical Form Liquid
Quantity 1 L
Packaging Glass Bottle
Grade Karl Fischer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.