Learn More
Description
- One component reagent for volumetric Karl Fischer titration for aldehydes and ketones
- 1 mL ≙ ca. 5 mg H2O Aquastar™
Specifications
Specifications
| Chemical Name or Material | CombiTitrant 5 Keto |
| Quantity | 1 L |
| Packaging | Glass Bottle |
| Grade | Karl Fischer |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.