Learn More
Research Products International Corp BEAKERplus COMBO Beaker and Flask, 150 ml, 6 pack
SDP
Catalog No. 502229469
Encompass
BEAKERplus is made of heavy duty borosilicate glass that withstands high temperatures. Features a wide rim with pour spout and accommodates funnels or filters. Narrow constricted neck provides extra protection from liquid splash during agitation. Can be autoclaved.
Catalog No. 50-222-9469 Supplier Research Products International Corp Supplier No. 164120
BEAKERplus is made of heavy duty borosilicate glass that withstands high temperatures. Features a wide rim with pour spout and accommodates funnels or filters. Narrow constricted neck provides extra protection from liquid splash during agitation. Can be autoclaved.