Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC COMBO EYE/FACE WASH/SHWER

Catalog No. 5000788601
Encompass

This drench shower and eye/face wash station provides emergency decontamination with a 1-1/4 in inlet and a 20 GPM shower flow. It features a bowl, triangular pull rod, and push flag activated mechanisms, a galvanized steel pipe, and a plastic shower head. The unit offers a 3 GPM bowl flow and is corrosion resistant, with options for top or mid supply.

  • Drench shower with eye and face wash station
  • Stainless steel bowl material
  • Push flag activation method
  • Plastic shower head material
  • Galvanized steel pipe material
  • Open bowl wash station style
  • Bowl flow pressure set at 30 psi
  • Bowl flow rate of 3 GPM
  • Shower flow pressure set at 30 psi
  • Shower flow rate of 20 GPM
  • Complies with ANSI Z358.1-2004
  • Manufactured in the United States

Catalog No. 50-007-88601 Supplier MSC Supplier No. 86205507
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This drench shower and eye/face wash station provides emergency decontamination with a 1-1/4 in inlet and a 20 GPM shower flow. It features a bowl, triangular pull rod, and push flag activated mechanisms, a galvanized steel pipe, and a plastic shower head. The unit offers a 3 GPM bowl flow and is corrosion resistant, with options for top or mid supply.

  • Drench shower with eye and face wash station
  • Stainless steel bowl material
  • Push flag activation method
  • Plastic shower head material
  • Galvanized steel pipe material
  • Open bowl wash station style
  • Bowl flow pressure set at 30 psi
  • Bowl flow rate of 3 GPM
  • Shower flow pressure set at 30 psi
  • Shower flow rate of 20 GPM
  • Complies with ANSI Z358.1-2004
  • Manufactured in the United States

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.