Learn More
Research Products International Corp Combination Head for One Microplate or Microtubes, for use with BenchMixer Vortex Mixer
SDP
Catalog No. 501367758
Encompass
Combination Head for One Microplate and Microtubes is an optional part for use with BenchMixer Vortex Mixer. Holds 66 tubes (38 x 1.5ml and 28 x 0.5ml) or one microplate.
Catalog No. 50-136-7758 Supplier Research Products International Corp Supplier No. 400812
Combination Head for One Microplate and Microtubes is an optional part for use with BenchMixer Vortex Mixer. Holds 66 tubes (38 x 1.5ml and 28 x 0.5ml) or one microplate.