Learn More
Saf T Pak Inc STP-740 - Disposable 2 - Part Secondary Pressure Vessel (95 kPa), Extra Large
SDP
Catalog No. NC9259569 Shop All Saf-T-Pak Inc. Products
Encompass_Preferred
Polybag and Envelope, Large inner leakproof poly bag (14 x 19in.), Tyvek outer envelope (12 x 16in.), Pressure tested to hold 95kPa, For transport of forensic evidence, Body parts, Diagnostic kits, Use with non-infectious specimens, 50/Case
Catalog No. NC9259569 Supplier Saf T Pak Inc Supplier No. STP740
Polybag and Envelope, Large inner leakproof poly bag (14 x 19in.), Tyvek outer envelope (12 x 16in.), Pressure tested to hold 95kPa, For transport of forensic evidence, Body parts, Diagnostic kits, Use with non-infectious specimens, 50/Case