Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Veritor™ Combo Pack: 2 SARS/Flu Kits and 1 Analyzer

Catalog No. B256095 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
60 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B256095 60 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Available for pre-order, orders will begin shipping August 15th
Catalog No. B256095 Supplier BD Veritor™ Supplier No. 256095
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Available for pre-order, orders will begin shipping August 15th

Promotional kit for rapid detection and differentiation of SARS-CoV-2 and Influenza

  • Detect and differentiate between SARS-CoV-2, Flu A, and Flu B from one sample on one assay in just 15 minutes
  • Identical workflow to the SARS-CoV-2 assay Dual anterior nares sample collection
  • Clear, digitally displayed results that detect positives not recognized by the unaided eye
  • Virtual SKU contains QTY 2 of BD Veritor™ System for Rapid Detection of SARS-CoV-2 & Flu A+B (256088) and QTY 1 of BD Veritor™ Plus Analyzer (256066)
  • Contains 60 tests and 1 instrument
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Certificate of Conformance
Content And Storage 2°C to 30°C
Control Sets SARS+, Flu A+, Flu B+ included
CPT Code 87428
CE Marker No
Clia Complexity Waived
Detectable Analytes Influenza A, Influenza B, SARS-CoV-2
Sample Type Dual Anterior Nares
Sensitivity 86.7%
Type Rapid Antigen
For Use With (Application) Lateral Flow Immunoassay
For Use With (Equipment) BD Veritor Plus Analyzer (256066) Included in This Promotional Pk.
Quantity 60 Tests
Specificity 0.995
Test Time 15 min.
Form Liquid
Show More Show Less

*The BD Veritor™ System for Rapid Detection of SARS-CoV-2 & Flu A+B is intended for the in vitro, simultaneous qualitative detection and differentiation of SARS-CoV-2 nucleocapsid antigen and/or influenza A and B nucleoprotein antigens directly from anterior nasal swab samples taken from individuals who are suspected of a viral respiratory infection consistent with COVID-19 by a healthcare provider, within the first six days of symptom onset.

  • This product has not been FDA cleared or approved, but has been authorized by FDA under an EUA for use by authorized laboratories.
  • This product has been authorized only for the detection of proteins from SARS-CoV-2, influenza A and influenza B, not for any other viruses or pathogens.
  • The emergency use of this product is only authorized for the duration of the declaration that circumstances exist justifying the authorization of emergency use of in vitro diagnostics for detection and/or diagnosis of COVID-19 under Section 564(b)(1) of the Federal Food, Drug and Cosmetic Act, 21 U.S.C. § 360bbb-3(b)(1), unless the declaration is terminated or authorization is revoked sooner.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.