Learn More
Cayman Chemical Combo Precelys EvolutIon 1ea
SDP
Catalog No. 502917930
Encompass_Preferred
The Precellys Evolution homogenizer and Cryolys Evolution cooling option designed to grind or homogenize samples under dry ice or liquid nitrogen cooled air
Catalog No. 50-291-7930 Supplier Cayman Chemical Supplier No. 387081 EA
Add to Cart
Add to Cart
Request a Quote
The Precellys Evolution homogenizer and Cryolys Evolution cooling option designed to grind or homogenize samples under dry ice or liquid nitrogen cooled air