Learn More
Cooper Atkins Corporation COMBO PROBE 36 LONG
Catalog No. NC0403416
Encompass_Preferred
Large Heavy Duty T-handle designed for processing applications such as Kettles or Vats. Temp Range: 100 deg.F to 500 deg.F (-73 deg.C to 260 deg.C). Shaft (L)35 in., Cord (L)36 in., Type-K Thermocouple connection
Catalog No. NC0403416 Supplier Cooper Atkins Corporation Supplier No. 50701K
Large Heavy Duty T-handle designed for processing applications such as Kettles or Vats. Temp Range: 100 deg.F to 500 deg.F (-73 deg.C to 260 deg.C). Shaft (L)35 in., Cord (L)36 in., Type-K Thermocouple connection