Learn More
PHCbi 11.5 cu.ft. Ref and 4.8 cu.ft. Frzr Pharmaceutical Refrigerator Freezer Combo
Catalog No. 502720039
Encompass_Preferred
The MPR-N450FSH-PA Pharmaceutical Refrigerator with Freezer Combo provides a storage environment with precise temperature control and superior uniformity ideal for biologic material, vaccine and pharmaceuticals. This unique combination refrigerator and freezer in one cabinet provides for safe storage of all stored product in a small single footprint, conserving lab space.Effective capacity: Refrigerator 11.5 cu.ft. (326 L) | Freezer 4.8 cu.ft. (136 L)Temperature range: Refrigerator 2℃ to 14℃ | Freezer -20℃ to -30℃
Catalog No. 50-272-0039 Supplier PHCbi Supplier No. MPRN450FSHPA
Add to Cart
Add to Cart
Request a Quote
The MPR-N450FSH-PA Pharmaceutical Refrigerator with Freezer Combo provides a storage environment with precise temperature control and superior uniformity ideal for biologic material, vaccine and pharmaceuticals. This unique combination refrigerator and freezer in one cabinet provides for safe storage of all stored product in a small single footprint, conserving lab space.Effective capacity: Refrigerator 11.5 cu.ft. (326 L) | Freezer 4.8 cu.ft. (136 L)Temperature range: Refrigerator 2℃ to 14℃ | Freezer -20℃ to -30℃