Learn More
PHCbi Combination refrigerator/freezer
Catalog No. 502720041
Encompass_Preferred
PHCbi brand 6.3 cu.ft ( 179 L) refrigerator and 2.8cu.ft (80 L) freezer combination unit with separate refrigeration systems. These purpose-built combination units are designed for temperature uniformity not commonly found in commerical cabinets.Temperature RangeRefrigerator 2℃ to 14℃ | Freezer -20℃ to -30℃
Catalog No. 50-272-0041 Supplier PHCbi Supplier No. MPRN250FSHPA
Add to Cart
Add to Cart
Request a Quote
PHCbi brand 6.3 cu.ft ( 179 L) refrigerator and 2.8cu.ft (80 L) freezer combination unit with separate refrigeration systems. These purpose-built combination units are designed for temperature uniformity not commonly found in commerical cabinets.Temperature RangeRefrigerator 2℃ to 14℃ | Freezer -20℃ to -30℃