Learn More
Magbio Genomics Inc COMBO SEPARATION RK 3X3
SDP
Catalog No. NC2003782
Encompass
Combo Magnetic Separation Rack is designed to simplify the manual processing of magnetic bead separations for nucleic acid purification and clean up. With the Combo Magnetic Separation Rack one can process either 3 x 50 ml volume conical tubes with DNA or RNA samples or 3 x 15 ml volume conical tubes with nucleic acids. The use of magnetic separation devices is essential in any paramagnetic bead-based DNA and RNA purification process. MAGBIO GENOMICS INC mfg# MBMS-31550
Catalog No. NC2003782 Supplier Magbio Genomics Inc Supplier No. MBMS31550
Combo Magnetic Separation Rack is designed to simplify the manual processing of magnetic bead separations for nucleic acid purification and clean up. With the Combo Magnetic Separation Rack one can process either 3 x 50 ml volume conical tubes with DNA or RNA samples or 3 x 15 ml volume conical tubes with nucleic acids. The use of magnetic separation devices is essential in any paramagnetic bead-based DNA and RNA purification process. MAGBIO GENOMICS INC mfg# MBMS-31550