Learn More
Poltex Combo Small/Large Labwipe Holder
SDP
Catalog No. 22000282
Change view
Click to view available options
Mounting:
Magnetic Mount
Wall Mount
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
22-000-282 Wall Mount
22-000-283 Magnetic Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-000-282 Supplier Poltex Supplier No. COMBOLABW
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Poltex Combo Small/Large Labwipe Holder holds one box each of large and small laboratory wipes
  • Durable PETG construction
  • Mounts horizontally or vertically
Specifications
Product Type Labwipe holder
Color Clear
Dimensions (L x W x H) 3.75 x 5 x 20 in.
Mounting Wall Mount
Quantity 1 Ea.