Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Combo Veritor Plus + CLIA RSV

Catalog No. B256128 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
CE Marker:
Yes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. CE Marker
B256128 Yes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B256128 Supplier BD Supplier No. 256128
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This package comes with two RSV Kits for Respiratory Syncytial Virus and 1 BD Veritor PLUS Analyzer Software Version 5.5.

  • Qty 2 BD Veritor RSV Physician Kits (30 tests/kit) + 1 BD Veritor Analyzer.
TRUSTED_SUSTAINABILITY

Specifications

CE Marker Yes
Clia Complexity Waived
Detectable Analytes Respiratory Syncytial Virus
Format Kit
Sample Type Nasal Swab
Sensitivity High
Type RSV Kits
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.