Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One ComboPlate™ 24 Well Protein Crystallization Plate

Catalog No. 07001011 Shop All Greiner Bio-One Products
Change view
Click to view available options
Well Shape:
Flat Bottom (F-well)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Well Shape
07-001-011 Flat Bottom (F-well)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-001-011 Supplier Greiner Bio-One Supplier No. 66215045

missing translation for 'validMainframeMsg'

Used for hanging drop application.

  • Universal design
  • Fattened raised ring around each well to reduce risk of cross-contamination
  • Possible to seal wells with silicone grease and cover slips
  • Slightly raised lid to protect during transportation
TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid With Lid
Array 4 x 6
Color Clear
Material Polystyrene
No. of Wells 24
Sterility Non-sterile
Total Volume (Metric) Well 3 mL
Well Shape Flat Bottom (F-well)
For Use With (Application) Protein crystallization
Includes Crystal bridges (Pre inserted, 24)
Well Design Flattened, raised ring
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.