Learn More
Greiner Bio-One ComboPlate™ 24-well Protein Crystallization Plate, Pre-greased
Catalog No. 07000669 Shop All Greiner Bio-One Products
Change view
Click to view available options
Well Shape:
Flat Bottom (F-well)
Catalog No. Well Shape
07-000-669 Flat Bottom (F-well)
1 options
Catalog No. 07-000-669 Supplier Greiner Bio-One Supplier No. 662050
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Ideal for protein crystallization. Greiner Bio-One ComboPlate™ 24-Well Protein Crystallization Plate, Pre-greased is universal plate with lid for crystallography.
  • Virgin, high-clarity polystyrene combined with distortion-free bottom provides optical properties for microscopic inspection of crystal growth
  • Flattened raised ring around each well reduces risk of cross-contamination
  • Raised protection lid protects cover slips and sealer during transportation and storage
  • Flat well bottom
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid With Lid
Array 4 x 6
Packaging Inner bags of 6
Color Clear
Material Polystyrene
Volume (Metric) Well 3300 μL
No. of Wells 24
Sterility Non-sterile
Surface Treatment Pre-greased
Well Shape Flat Bottom (F-well)
No. per Case 24
Quantity 24/Cs.
Height (Metric) 18.8 mm
Outside Diameter (Metric) Well: 16.3 mm