Learn More
Medchemexpress LLC Combretastatin A-1 | 109971-63-3 | 97.0% | 10 MG
SDP
Catalog No. 5000230730
Encompass_Preferred
Combretastatin A-1 is a microtubule polymerization inhibitor used as a research standard to study tubulin dynamics, signal transduction, and anti-tumor activity. It binds the colchicine-binding site of tubulin and has documented anti-vascular effects in biological studies.
  • Microtubule polymerization inhibitor that targets the colchicine-binding site.
  • Used for research into tubulin dynamics and Wnt/β-catenin signaling.
  • Reported anti-tumor and anti-vascular biological activity.
  • Provided as a 10 mg research standard suitable for analytical and biological assays.
Catalog No. 50-002-30730 Supplier Medchemexpress LLC Supplier No. HY12199310MG
Combretastatin A-1 is a microtubule polymerization inhibitor used as a research standard to study tubulin dynamics, signal transduction, and anti-tumor activity. It binds the colchicine-binding site of tubulin and has documented anti-vascular effects in biological studies.
  • Microtubule polymerization inhibitor that targets the colchicine-binding site.
  • Used for research into tubulin dynamics and Wnt/β-catenin signaling.
  • Reported anti-tumor and anti-vascular biological activity.
  • Provided as a 10 mg research standard suitable for analytical and biological assays.