Learn More
Medchemexpress LLC Combretastatin A-1 | 109971-63-3 | 97.0% | 100 MG
SDP
Catalog No. 5000230729
Encompass_Preferred
Combretastatin A-1 (CAS 109971-63-3) is a microtubule polymerization inhibitor supplied as a research-grade solid. It is used as an analytical standard and reagent for studying tubulin dynamics, cell biology assays, and biochemical experiments. Common handling involves dissolution in DMSO to prepare concentrated stock solutions for in vitro use.
  • Microtubule polymerization inhibitor for tubulin-related studies.
  • Suitable as an analytical standard and research reagent.
  • High purity appropriate for biochemical and cell-based assays.
  • Stable as a powder when stored under recommended frozen conditions.
  • Soluble in DMSO for preparation of concentrated stock solutions.
Catalog No. 50-002-30729 Supplier Medchemexpress LLC Supplier No. HY121993100MG
Combretastatin A-1 (CAS 109971-63-3) is a microtubule polymerization inhibitor supplied as a research-grade solid. It is used as an analytical standard and reagent for studying tubulin dynamics, cell biology assays, and biochemical experiments. Common handling involves dissolution in DMSO to prepare concentrated stock solutions for in vitro use.
  • Microtubule polymerization inhibitor for tubulin-related studies.
  • Suitable as an analytical standard and research reagent.
  • High purity appropriate for biochemical and cell-based assays.
  • Stable as a powder when stored under recommended frozen conditions.
  • Soluble in DMSO for preparation of concentrated stock solutions.