Learn More
Medchemexpress LLC Combretastatin A-1 | 109971-63-3 | 97.0% | 25 MG
SDP
Catalog No. 5000230600
Encompass_Preferred
Combretastatin A-1 is a small-molecule microtubule polymerization inhibitor that binds the colchicine-binding site on tubulin, leading to tubulin depolymerization, AKT deactivation, and inhibition of the Wnt/β-catenin signaling pathway. Supplied for research use only.
  • Microtubule polymerization inhibitor that targets the colchicine-binding site on tubulin.
  • Inhibits Wnt/β-catenin signaling via tubulin depolymerization-mediated AKT deactivation.
  • High purity (97.0%) suitable for research and analytical applications.
  • Available as solid and DMSO solution formats, including 25 MG solid pack sizes.
  • Molecular formula C18H20O6 and molecular weight 332.35 g·mol⁻1.
Catalog No. 50-002-30600 Supplier Medchemexpress LLC Supplier No. HY12199325MG
Combretastatin A-1 is a small-molecule microtubule polymerization inhibitor that binds the colchicine-binding site on tubulin, leading to tubulin depolymerization, AKT deactivation, and inhibition of the Wnt/β-catenin signaling pathway. Supplied for research use only.
  • Microtubule polymerization inhibitor that targets the colchicine-binding site on tubulin.
  • Inhibits Wnt/β-catenin signaling via tubulin depolymerization-mediated AKT deactivation.
  • High purity (97.0%) suitable for research and analytical applications.
  • Available as solid and DMSO solution formats, including 25 MG solid pack sizes.
  • Molecular formula C18H20O6 and molecular weight 332.35 g·mol⁻1.