Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Combretastatin A-1 | 109971-63-3 | 97.0% | 5 MG
SDP

Catalog No. 5000231610
Encompass_Preferred

Combretastatin A-1 is a microtubule polymerization inhibitor supplied as a high-purity research standard for in vitro studies. It binds the colchicine-binding site on tubulin and is commonly used to investigate microtubule dynamics and anticancer mechanisms.

  • Purity 97.0%.
  • Molecular weight 332.35 g/mol.
  • Chemical formula C18H20O6.
  • Supplied as a 5 mg solid.
  • Suitable for research and analytical applications.
  • Documentation available: datasheet, certificate of analysis, SDS, and RP-HPLC report.

Catalog No. 50-002-31610 Supplier Medchemexpress LLC Supplier No. HY1219935MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Combretastatin A-1 is a microtubule polymerization inhibitor supplied as a high-purity research standard for in vitro studies. It binds the colchicine-binding site on tubulin and is commonly used to investigate microtubule dynamics and anticancer mechanisms.

  • Purity 97.0%.
  • Molecular weight 332.35 g/mol.
  • Chemical formula C18H20O6.
  • Supplied as a 5 mg solid.
  • Suitable for research and analytical applications.
  • Documentation available: datasheet, certificate of analysis, SDS, and RP-HPLC report.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.