Learn More
Medchemexpress LLC Combretastatin A-1 | 109971-63-3 | >98.0% | 50 MG
SDP
Catalog No. 5000230534
Encompass_Preferred
Combretastatin A-1 is a cis-stilbene microtubule polymerization inhibitor (CAS 109971-63-3) used in research to study tubulin dynamics and as an experimental antitumor agent. It is supplied as a crystalline solid for research use and is typically handled and stored under cold conditions to preserve stability.
  • Microtubule polymerization inhibitor that binds the colchicine site on tubulin.
  • Used for cellular assays and in vivo cancer research models.
  • Crystalline solid; typically stored at -20°C for long-term stability.
  • Molecular weight 332.4 g/mol, chemical formula C18H20O6.
  • Provided in research-scale quantities suitable for assay development and preclinical studies.
Catalog No. 50-002-30534 Supplier Medchemexpress LLC Supplier No. HY12199350MG
Combretastatin A-1 is a cis-stilbene microtubule polymerization inhibitor (CAS 109971-63-3) used in research to study tubulin dynamics and as an experimental antitumor agent. It is supplied as a crystalline solid for research use and is typically handled and stored under cold conditions to preserve stability.
  • Microtubule polymerization inhibitor that binds the colchicine site on tubulin.
  • Used for cellular assays and in vivo cancer research models.
  • Crystalline solid; typically stored at -20°C for long-term stability.
  • Molecular weight 332.4 g/mol, chemical formula C18H20O6.
  • Provided in research-scale quantities suitable for assay development and preclinical studies.